Thermo Scientific Chemicals

1-Chloro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A15403.30
250 g, Each
Thermo Scientific Chemicals

1-Chloro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A15403.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15403.30
also known as A15403-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-73-3
IUPAC Name
1-chloro-2-nitrobenzene
Molecular Formula
C6H4ClNO2
InChI Key
BFCFYVKQTRLZHA-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC=C1Cl
Appearance (Color)
Yellow
Melting Point (clear melt)
30.0-36.0?C
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.5%

Description

1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

Solubility
Soluble in water 0.59 g/L.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text