Thermo Scientific Chemicals

DL-2,4-Diaminobutyric acid dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A15408.06
5 g, Each
Thermo Scientific Chemicals

DL-2,4-Diaminobutyric acid dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A15408.06
5 g, Each
Quantity
Catalog number: A15408.06
also known as A15408-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65427-54-5
IUPAC Name
(2R)-2,4-diazaniumylbutanoate
Molecular Formula
C4H11N2O2
InChI Key
OGNSCSPNOLGXSM-GSVOUGTGSA-O
SMILES
[NH3+]CC[C@@H]([NH3+])C([O-])=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%

Description

2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activityagainst mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activity against mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

Solubility
Soluble in water 50 mg/mL.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text