Pyridine-2-carboxamide, 98%, Thermo Scientific Chemicals
Pyridine-2-carboxamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine-2-carboxamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A15410.14
also known as A15410-14
Price (USD)/ Each
196.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
196.00
Add to cart
Pyridine-2-carboxamide, 98%, Thermo Scientific Chemicals
Catalog numberA15410.14
Price (USD)/ Each
196.00
-
Add to cart
Chemical Identifiers
CAS1452-77-3
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)103.0-110.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only