Thermo Scientific Chemicals

Terephthalaldehyde mono(diethyl acetal), 97%, stab., Thermo Scientific Chemicals

Catalog number: A15413.14
25 g, Each
Thermo Scientific Chemicals

Terephthalaldehyde mono(diethyl acetal), 97%, stab., Thermo Scientific Chemicals

Catalog number: A15413.14
25 g, Each
Quantity
Catalog number: A15413.14
also known as A15413-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81172-89-6
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%
Refractive Index
1.5050-1.5090 @ 20?C
Form
Liquid
Stabilizer
0.1% triethylamine

Description

4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text