Thermo Scientific™

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A15415.18
50 g, Each
Thermo Scientific™

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A15415.18
50 g, Each
Quantity
Catalog number: A15415.18
also known as A15415-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6276-54-6
IUPAC Name
hydrogen 3-chloropropan-1-amine chloride
Molecular Formula
C3H9Cl2N
InChI Key
IHPRVZKJZGXTBQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCCCl
Form
Crystalline powder and/or chunks
Assay (Titration ex Chloride)
≥97.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White to cream to yellow to pale brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text