5,6-Dimethyl-2-benzimidazolinone, 98%, Thermo Scientific Chemicals
5,6-Dimethyl-2-benzimidazolinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,6-Dimethyl-2-benzimidazolinone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A15421.03
also known as A15421-03
Price (USD)/ Each
69.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
69.00
Add to cart
5,6-Dimethyl-2-benzimidazolinone, 98%, Thermo Scientific Chemicals
Catalog numberA15421.03
Price (USD)/ Each
69.00
-
Add to cart
Chemical Identifiers
CAS2033-30-9
IUPAC Name5,6-dimethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular FormulaC9H10N2O
InChI KeyORWJLFLEIZBRBR-UHFFFAOYSA-N
SMILESCC1=C(C)C=C2NC(=O)NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Pale brown to brown
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only