3-(1-Naphthyl)acrylic acid, 98%, Thermo Scientific Chemicals
3-(1-Naphthyl)acrylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(1-Naphthyl)acrylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15422.09
also known as A15422-09
Price (USD)/ Each
66.70
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
66.70
Add to cart
3-(1-Naphthyl)acrylic acid, 98%, Thermo Scientific Chemicals
Catalog numberA15422.09
Price (USD)/ Each
66.70
-
Add to cart
Chemical Identifiers
CAS13026-12-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)208.0-215.0?C
Assay (Silylated GC)≥97.5%
FormPowder
3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only