Thermo Scientific Chemicals

1H-Benzotriazole, 99%, Thermo Scientific Chemicals

Catalog number: A15423.0B
1000 g, Each
Thermo Scientific Chemicals

1H-Benzotriazole, 99%, Thermo Scientific Chemicals

Catalog number: A15423.0B
1000 g, Each
Quantity
Catalog number: A15423.0B
also known as A15423-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-14-7
IUPAC Name
2H-1,2,3-benzotriazole
Molecular Formula
C6H5N3
InChI Key
QRUDEWIWKLJBPS-UHFFFAOYSA-N
SMILES
N1N=C2C=CC=CC2=N1
Assay from Supplier's CofA
≥98.5% (GC, US sourced material)
Melting Point (clear melt)
94-100?C
Form
Powder or crystals or granules or needles or flakes
Assay (GC)
≥98.5% (UK sourced material)
Appearance (Color)
White to cream to yellow to pale brown

Description

1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

Solubility
Soluble in water, benzene, toluene, chloroform, ethanol and N,N-dimethylformamide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and heavy metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text