4-Methylthiazole, 99%, Thermo Scientific Chemicals
4-Methylthiazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylthiazole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A15428.22
also known as A15428-22
Price (USD)/ Each
211.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
211.00
Add to cart
4-Methylthiazole, 99%, Thermo Scientific Chemicals
Catalog numberA15428.22
Price (USD)/ Each
211.00
-
Add to cart
Specifications
Chemical Name or Material4-Methylthiazole
CAS693-95-8
Health Hazard 1H226-H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only