4-Cyanobenzoic acid, 98%, Thermo Scientific Chemicals
4-Cyanobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A15434.14
also known as A15434-14
Price (USD)/ Each
132.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
132.00
Add to cart
4-Cyanobenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberA15434.14
Price (USD)/ Each
132.00
-
Add to cart
Chemical Identifiers
CAS619-65-8
IUPAC Name4-cyanobenzoic acid
Molecular FormulaC8H5NO2
InChI KeyADCUEPOHPCPMCE-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder or crystalline powder
Assay (Silylated GC)≥97.5%
Appearance (Color)White to cream to yellow or pale brown
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases.
RUO – Research Use Only

General References:

  1. Sun Hee Kim; Sang Jung Ahn; and Kwan Kim. Vibrational Spectroscopic Study of 4-Cyanobenzoic Acid Adsorbed on Silver.J. Phys. Chem.1996, 100 (17), 7174-7180.
  2. Yan Li; Fa-Kun Zheng; Xi Liu; Wen-Qiang Zou; Guo-Cong; Can-Zhong Lu and Jin-Shun Huang. Crystal Structures and Magnetic and Luminescent Properties of a Series of Homodinuclear Lanthanide Complexes with 4-Cyanobenzoic Ligand.Inorg. Chem.2006, 45 (16), 6308-6316.