Thermo Scientific Chemicals

2-Cyclohexen-1-one, 97%, Thermo Scientific Chemicals

Catalog number: A15435.14
25 g, Each
Thermo Scientific Chemicals

2-Cyclohexen-1-one, 97%, Thermo Scientific Chemicals

Catalog number: A15435.14
25 g, Each
Quantity
Catalog number: A15435.14
also known as A15435-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
930-68-7
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4855-1.4905 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%

Description

2-Cyclohexen-1-one is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclohexen-1-one is used as intermediates.

Solubility
Soluble in water, alcohols, benzene and acetone.

Notes
Store away from oxidizing agents and strong bases. Incompatible with reducing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text