7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals
7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15441.06
also known as A15441-06
Price (USD)/ Each
38.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.70
Add to cart
7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals
Catalog numberA15441.06
Price (USD)/ Each
38.70
-
Add to cart
Chemical Identifiers
CAS531-59-9
IUPAC Name7-methoxy-2H-chromen-2-one
Molecular FormulaC10H8O3
InChI KeyLIIALPBMIOVAHH-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=CC(=O)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale orange to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)115.0-122.0?C
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. R.K. Reen; S. Ramakanth; F.J. Wiebel; M.P. Jain∗, J. Singh. Dealkylation of 7-methoxycoumarin as assay for measuring constitutive and phenobarbital-inducible cytochrome P450s. Analytical Biochemistry. 1991, 194 (2), 243-249.
  2. K. Gnanaguru; N. Ramasubbu; K. Venkatesan; V. Ramamurthy. Topochemical solid state photodimerization of non-ideally oriented monomers: 7-chlorocoumarin and 7-methoxycoumarin. Journal of Photochemistry. 1984, 27 (3), 355-362.