Thermo Scientific Chemicals

7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals

Catalog number: A15441.22
100 g, Each
Thermo Scientific Chemicals

7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals

Catalog number: A15441.22
100 g, Each
Quantity
Catalog number: A15441.22
also known as A15441-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
531-59-9
IUPAC Name
7-methoxy-2H-chromen-2-one
Molecular Formula
C10H8O3
InChI Key
LIIALPBMIOVAHH-UHFFFAOYSA-N
SMILES
COC1=CC=C2C=CC(=O)OC2=C1
Appearance (Color)
White to cream or pale orange to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
115.0-122.0?C

Description

7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text