Thermo Scientific Chemicals

2,5-Dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog number: A15442.18
50 g, Each
Thermo Scientific Chemicals

2,5-Dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog number: A15442.18
50 g, Each
Quantity
Catalog number: A15442.18
also known as A15442-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16110-09-1
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
56.5-62.5?C

Description

2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text