Cyanuric acid, 99%, Thermo Scientific Chemicals
Cyanuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyanuric acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1000 g
Catalog number A15447.30
also known as A15447-30
Price (USD)/ Each
58.65
Online exclusive
65.00 
Save 6.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.00 
Save 6.35 (10%)
Add to cart
Cyanuric acid, 99%, Thermo Scientific Chemicals
Catalog numberA15447.30
Price (USD)/ Each
58.65
Online exclusive
65.00 
Save 6.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-80-5
IUPAC Name1,3,5-triazinane-2,4,6-trione
Molecular FormulaC3H3N3O3
InChI KeyZFSLODLOARCGLH-UHFFFAOYSA-N
SMILESO=C1NC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5% to ≤101.5%
Assay (Silylated GC)≥98.5%
Identification (FTIR)Conforms
FormPowder
Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Murphy, J. L.; Arrowood, M. J.; Lu, X.; Hlavsa, M. C.; Beach, M. J.; Hill, V. R. Effect of Cyanuric Acid on the Inactivation of Cryptosporidium parvum under Hyperchlorination Conditions. Environ. Sci. Technol. 2015, 49 (12), 7348-7355.
  2. Reva, I. Comment on Density functional theory studies on molecular structure, vibrational spectra and electronic properties of cyanuric acid. Spectrochim. Acta, Part A 2015, 151, 232-236.