Thermo Scientific Chemicals

Bromocyclopentane, 98+%, Thermo Scientific Chemicals

Catalog number: A15455.22
100 g, Each
Thermo Scientific Chemicals

Bromocyclopentane, 98+%, Thermo Scientific Chemicals

Catalog number: A15455.22
100 g, Each
Quantity
Catalog number: A15455.22
also known as A15455-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-43-9
IUPAC Name
bromocyclopentane
Molecular Formula
C5H9Br
InChI Key
BRTFVKHPEHKBQF-UHFFFAOYSA-N
SMILES
BrC1CCCC1
Form
Liquid
Comment
May darken on storage
Refractive Index
1.4870-1.4910 @ 20?C
Appearance (Color)
Clear colorless to yellow to brown
Identification (FTIR)
Conforms

Description

Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text