Thermo Scientific Chemicals

Diiodomethane, 99%, stab., Thermo Scientific Chemicals

Catalog number: A15457.22
100 g, Each
Thermo Scientific Chemicals

Diiodomethane, 99%, stab., Thermo Scientific Chemicals

Catalog number: A15457.22
100 g, Each
Quantity
Catalog number: A15457.22
also known as A15457-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-11-6
IUPAC Name
diiodomethane
Molecular Formula
CH2I2
InChI Key
NZZFYRREKKOMAT-UHFFFAOYSA-N
SMILES
ICI
Appearance (Color)
Clear colorless to yellow to red or orange-brown
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Stabilizer
Copper wire

Description

Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with alkali metal salts, strong oxidizing agents, strong bases, metals, lithium, potassium, sodium, sodium oxides and alloys.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text