3-Dimethylaminopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
3-Dimethylaminopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Dimethylaminopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A15459.0E
also known as A15459-0E
Price (USD)/ Each
451.65
Online exclusive
502.00 
Save 50.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
451.65
Online exclusive
502.00 
Save 50.35 (10%)
Add to cart
3-Dimethylaminopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberA15459.0E
Price (USD)/ Each
451.65
Online exclusive
502.00 
Save 50.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5407-04-5
IUPAC Namehydrogen (3-chloropropyl)dimethylamine chloride
Molecular FormulaC5H13Cl2N
InChI KeyLJQNMDZRCXJETK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
Melting Point (clear melt)136.0-144.0°C
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only