Di-n-heptyl phthalate, 98+%, Thermo Scientific Chemicals
Di-n-heptyl phthalate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-heptyl phthalate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A15466.09
also known as A15466-09
Price (USD)/ Each
73.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
73.50
Add to cart
Di-n-heptyl phthalate, 98+%, Thermo Scientific Chemicals
Catalog numberA15466.09
Price (USD)/ Each
73.50
-
Add to cart
Chemical Identifiers
CAS3648-21-3
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4835-1.4875 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
FormLiquid
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

Solubility
Sparingly soluble in water.

Notes
Incompatible with Strong acids, strong bases,.
RUO – Research Use Only

General References:

  1. Ian Cousins; Donald Mackay. Correlating the physical-chemical properties of phthalate esters using the three solubility approach.Chemosphere.2000, 41 1389-1399.