4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A15467.14
also known as A15467-14
Price (USD)/ Each
409.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
409.00
Add to cart
4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA15467.14
Price (USD)/ Each
409.00
-
Add to cart
Chemical Identifiers
CAS94108-56-2
IUPAC Name4-(trifluoromethoxy)benzene-1-sulfonyl chloride
Molecular FormulaC7H4ClF3O3S
InChI KeyUHCDBMIOLNKDHG-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4775-1.4815 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only