Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15470.14
also known as A15470-14
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
Catalog numberA15470.14
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10534-89-1
IUPAC Namecobalt(3+) hexaamine trichloride
Molecular FormulaCl3CoH18N6
InChI KeyJXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Appearance (Color)Orange
Identification (FTIR)Conforms
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

Solubility
Soluble in ammonia and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Howard, M. J.; Klemm, B. P.; Fierke, C. A. Mechanistic Studies Reveal Similar Catalytic Strategies for Phosphodiester Bond Hydrolysis by Protein-only and RNA-dependent Ribonuclease P. J. Biol. Chem. 2015, 290 (21), 13454-13464.
  2. Yao, Q.; Lu, Z. H.; Wang, Y.; Chen, X.; Feng, G. Synergetic Catalysis of Non-noble Bimetallic Cu-Co Nanoparticles Embedded in SiO2 Nanospheres in Hydrolytic Dehydrogenation of Ammonia Borane. J. Phys. Chem. C 2015, 119 (25), 14167-14174.