Thermo Scientific Chemicals

3-Methoxybenzyl alcohol, 97+%, Thermo Scientific Chemicals

Catalog number: A15471.14
25 g, Each
Thermo Scientific Chemicals

3-Methoxybenzyl alcohol, 97+%, Thermo Scientific Chemicals

Catalog number: A15471.14
25 g, Each
Quantity
Catalog number: A15471.14
also known as A15471-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6971-51-3
IUPAC Name
(3-methoxyphenyl)methanol
Molecular Formula
C8H10O2
InChI Key
IIGNZLVHOZEOPV-UHFFFAOYSA-N
SMILES
COC1=CC=CC(CO)=C1
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow to pale brown
Form
Liquid
Assay (GC)
≥97.0%

Description

3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in ether, and alcohol. Insoluble in water.

Notes
Incompatible with Oxidizing agents, acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text