Thermo Scientific Chemicals

2,6-Dimethyl-4-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog number: A15472.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dimethyl-4-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog number: A15472.14
25 g, Each
Quantity
Catalog number: A15472.14
also known as A15472-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2423-71-4
IUPAC Name
2,6-dimethyl-4-nitrophenol
Molecular Formula
C8H9NO3
InChI Key
FNORUNUDZNWQFF-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1O)[N+]([O-])=O
Form
Powder
Appearance (Color)
Orange to brown
Assay (GC)
≥97.5%

Description

It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text