Thermo Scientific Chemicals

4-Methylbenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15479.36
500 g, Each
Thermo Scientific Chemicals

4-Methylbenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15479.36
500 g, Each
Quantity
Catalog number: A15479.36
also known as A15479-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
134-84-9
IUPAC Name
(4-methylphenyl)(phenyl)methanone
Molecular Formula
C14H12O
InChI Key
WXPWZZHELZEVPO-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid or flakes

Description

4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place. Store in a tightly closed container. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text