Thermo Scientific Chemicals

Pentafluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15490.06
5 g, Each
Thermo Scientific Chemicals

Pentafluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15490.06
5 g, Each
Quantity
Catalog number: A15490.06
also known as A15490-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
653-37-2
IUPAC Name
2,3,4,5,6-pentafluorobenzaldehyde
Molecular Formula
C7HF5O
InChI Key
QJXCFMJTJYCLFG-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C=O)C(F)=C1F
Free acid (titration)
≤2.5%
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4490-1.4530 (as melt)
Form
Low-melting fused solid or liquid as melt

Description

Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.

Solubility
Sparingly Soluble in water (0.027 g/L at 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, air, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text