2-Chloro-4-fluoroaniline, 97%, Thermo Scientific Chemicals
2-Chloro-4-fluoroaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-4-fluoroaniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A15492.22
also known as A15492-22
Price (USD)/ Each
426.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
426.00
Add to cart
2-Chloro-4-fluoroaniline, 97%, Thermo Scientific Chemicals
Catalog numberA15492.22
Price (USD)/ Each
426.00
-
Add to cart
Chemical Identifiers
CAS2106-02-7
IUPAC Name2-chloro-4-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyXRAKCYJTJGTSMM-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5515-1.5565 @ 20°C
Appearance (Color)Clear colorless to yellow to brown or pale pink to dark purple
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only