Tri-n-propyl orthoformate, 97%, Thermo Scientific Chemicals
Tri-n-propyl orthoformate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-propyl orthoformate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A15503.14
also known as A15503-14
Price (USD)/ Each
61.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
61.80
Add to cart
Tri-n-propyl orthoformate, 97%, Thermo Scientific Chemicals
Catalog numberA15503.14
Price (USD)/ Each
61.80
-
Add to cart
Chemical Identifiers
CAS621-76-1
IUPAC Name1-(dipropoxymethoxy)propane
Molecular FormulaC10H22O3
InChI KeyRWNXXQFJBALKAX-UHFFFAOYSA-N
SMILESCCCOC(OCCC)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4050-1.4090 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only