Thermo Scientific Chemicals

4-Chlorophenyl phosphorodichloridate, 98+%, Thermo Scientific Chemicals

Catalog number: A15507.14
25 g, Each
Thermo Scientific Chemicals

4-Chlorophenyl phosphorodichloridate, 98+%, Thermo Scientific Chemicals

Catalog number: A15507.14
25 g, Each
Quantity
Catalog number: A15507.14
also known as A15507-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
772-79-2
IUPAC Name
4-chlorophenyl chlorophosphonochloridate
Molecular Formula
C6H4Cl3O2P
InChI Key
CCZMQYGSXWZFKI-UHFFFAOYSA-N
SMILES
ClC1=CC=C(OP(Cl)(Cl)=O)C=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.5375-1.5415 @ 20?C
Form
Liquid

Description

4-Chlorophenyl phosphorodichloridate is used as phosphorylation agent. Reactant for preparation of antimicrobial agents, antioxidant agents, antitumor agents, nucleotide anti-viral prodrug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenyl phosphorodichloridate is used as phosphorylation agent. Reactant for preparation of antimicrobial agents, antioxidant agents, antitumor agents, nucleotide anti-viral prodrug.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, alcohols, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text