Thermo Scientific Chemicals

1,3-Dibromo-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals

Catalog number: A15510.22
100 g, Each
Thermo Scientific Chemicals

1,3-Dibromo-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals

Catalog number: A15510.22
100 g, Each
Quantity
Catalog number: A15510.22
also known as A15510-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-48-5
IUPAC Name
1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
Molecular Formula
C5H6Br2N2O2
InChI Key
VRLDVERQJMEPIF-UHFFFAOYSA-N
SMILES
CC1(C)N(Br)C(=O)N(Br)C1=O
Assay (Iodometric Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale yellow

Description

It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

Solubility
Soluble in Alcohol, Ether, Benzene. Insoluble in Water.

Notes
Moisture sensitive. Store away from oxidizing agents, reducing agents, flammable substances, organic materials, metal powders & moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text