Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.
Solubility
Soluble in water (78 mg/L 25°C).
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only
General References:
- Karuppiah Nagaraj; Subramanian Ambika; Shanmugasundaram Rajasri; Subramanian Sakthinathan; Sankaralingam Arunachalam. Synthesis, micellization behavior, antimicrobial and intercalative DNA binding of some novel surfactant copper(II) complexes containing modified phenanthroline ligands. Colloids and Surfaces B: Biointerfaces. 2014, 122, 151-157.
- Shuai Zhang; Qinfu Liu, Hongfei Cheng; Xiaoguang Li; Fangui Zeng; Ray L. Intercalation of dodecylamine into kaolinite and its layering structure investigated by molecular dynamics simulation. Journal of Colloid and Interface Science. 2014, 430, 345-350.