Phthalide, 98+%, Thermo Scientific Chemicals
Phthalide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A15516.22
also known as A15516-22
Price (USD)/ Each
34.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
34.70
Add to cart
Phthalide, 98+%, Thermo Scientific Chemicals
Catalog numberA15516.22
Price (USD)/ Each
34.70
-
Add to cart
Chemical Identifiers
CAS87-41-2
IUPAC Name1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC8H6O2
InChI KeyWNZQDUSMALZDQF-UHFFFAOYSA-N
SMILESO=C1OCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point70.5-76.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only