Thermo Scientific Chemicals

2,6-Difluoropyridine, 98+%, Thermo Scientific Chemicals

Catalog number: A15522.22
100 g, Each
Thermo Scientific Chemicals

2,6-Difluoropyridine, 98+%, Thermo Scientific Chemicals

Catalog number: A15522.22
100 g, Each
Quantity
Catalog number: A15522.22
also known as A15522-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1513-65-1
IUPAC Name
2,6-difluoropyridine
Molecular Formula
C5H3F2N
InChI Key
MBTGBRYMJKYYOE-UHFFFAOYSA-N
SMILES
FC1=CC=CC(F)=N1
Assay (GC)
≥98.0%
Refractive Index
1.4355-1.4395 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

Solubility
Not miscible or difficult to mix.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text