Thermo Scientific Chemicals

Pyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: A15534.06
5 g, Each
Thermo Scientific Chemicals

Pyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: A15534.06
5 g, Each
Quantity
Catalog number: A15534.06
also known as A15534-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
289-95-2
IUPAC Name
pyrimidine
Molecular Formula
C4H4N2
InChI Key
CZPWVGJYEJSRLH-UHFFFAOYSA-N
SMILES
C1=CN=CN=C1
Refractive Index
1.5020-1.5060 as melt
Assay (GC)
≥98.0%
Form
Low melting solid or clear liquid
Appearance (Color)
Clear colorless to yellow to orange

Description

Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text