Thermo Scientific Chemicals

Polyethylene oxide, M.W. >5,000,000, Thermo Scientific Chemicals

Catalog number: A15536.22
100 g, Each
Thermo Scientific Chemicals

Polyethylene oxide, M.W. >5,000,000, Thermo Scientific Chemicals

Catalog number: A15536.22
100 g, Each
Quantity
Catalog number: A15536.22
also known as A15536-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Appearance (Color)
White
Form
Powder/flake

Description

It is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text