Thermo Scientific Chemicals

Methyl phenylacetate, 99%, Thermo Scientific Chemicals

Catalog number: A15537.0I
5000 g, Each
Thermo Scientific Chemicals

Methyl phenylacetate, 99%, Thermo Scientific Chemicals

Catalog number: A15537.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15537.0I
also known as A15537-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-41-7
IUPAC Name
methyl 2-phenylacetate
Molecular Formula
C9H10O2
InChI Key
CRZQGDNQQAALAY-UHFFFAOYSA-N
SMILES
COC(=O)CC1=CC=CC=C1
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Refractive Index
1.5055-1.5085 @ 20?C
Form
Liquid

Description

Methyl phenylacetate is utilized for partition coefficient measurement experiments. It is mainly used in the flavor industry and in perfumes to impart honey scents. Further, it acts as a precursor to prepare synthetic perfumes. It acts as an acylating agent and involved in the enantioselective acylation of beta-lactam intermediate using penicillin G amidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text