Thermo Scientific Chemicals

Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A15540.0B
1000 g, Each
Thermo Scientific Chemicals

Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A15540.0B
1000 g, Each
Quantity
Catalog number: A15540.0B
also known as A15540-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13478-00-7
IUPAC Name
nickel(2+) hexahydrate dinitrate
Molecular Formula
H12N2NiO12
InChI Key
AOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Green
Assay (Redox Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid

Description

Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

Solubility
Very soluble in water. Soluble in ethanol.

Notes
As an oxidizer, it is incompatible with reducing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text