Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1000 g
5000 g
2.5 kg
Catalog number A15540.22
also known as A15540-22
Price (USD)/ Each
28.65
Online exclusive
31.70 
Save 3.05 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
31.70 
Save 3.05 (10%)
Add to cart
Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Catalog numberA15540.22
Price (USD)/ Each
28.65
Online exclusive
31.70 
Save 3.05 (10%)
-
Add to cart
Chemical Identifiers
CAS13478-00-7
IUPAC Namenickel(2+) hexahydrate dinitrate
Molecular FormulaH12N2NiO12
InChI KeyAOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid
Appearance (Color)Green
Assay (Redox Titration)≥97.5 to ≤102.5%
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

Solubility
Very soluble in water. Soluble in ethanol.

Notes
As an oxidizer, it is incompatible with reducing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Lee, S. M.; Ko, Y. N.; Choi, S. H.; Kim, J. H.; Kang, Y. C. Capacitive properties of reduced graphene oxide microspheres with uniformly dispersed nickel sulfide nanocrystals prepared by spray pyrolysis. Electrochim. Acta 2015, 167, 287-293.
  2. Klein, A.; Axmann, P.; Yada, C.; Wohlfahrt-Mehrens, M. Improving the cycling stability of Li2MnO3 by surface treatment. J. Power Sources 2015, 2, 302-307.
  3. Wang, W. N.; Itoh, Y.; Lenggoro, I. W.; Okuyama, K. Nickel and Nickel oxide Nanoparticles Prepared from Nickel nitrate hexahydrate by a Low Pressure Spray Pyrolysis. Mat. Sci. Eng. B-Solid 2004, 111 (1), 69-76.