Thermo Scientific Chemicals

2-Butynoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15542.06
5 g, Each
Thermo Scientific Chemicals

2-Butynoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15542.06
5 g, Each
Quantity
Catalog number: A15542.06
also known as A15542-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-93-2
IUPAC Name
but-2-ynoic acid
Molecular Formula
C4H4O2
InChI Key
LUEHNHVFDCZTGL-UHFFFAOYSA-N
SMILES
CC#CC(O)=O
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
73.0-80.0?C
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2-Butynoic acid is used as an intermediate in organic synthesis and pharmaceuticals. It is used as a synthon in various reactions, which includes cylcoalkylation of phenols to flavones and chromones. It is involved in the cyclization of gamma-butyrolactones. It is also used in the synthesis of Z-trisubstituted olefins through μ-alkylation. It plays an important role in the preparation of chirally pure 1,2,3,4-tetrahydroisoquinoline analogs, which finds application as anti-cancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butynoic acid is used as an intermediate in organic synthesis and pharmaceuticals. It is used as a synthon in various reactions, which includes cylcoalkylation of phenols to flavones and chromones. It is involved in the cyclization of gamma-butyrolactones. It is also used in the synthesis of Z-trisubstituted olefins through µ-alkylation. It plays an important role in the preparation of chirally pure 1,2,3,4-tetrahydroisoquinoline analogs, which finds application as anti-cancer agents.

Solubility
Soluble in water. Slightly soluble in methanol and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text