Thermo Scientific Chemicals

Chlorotriethylsilane, 98+%, Thermo Scientific Chemicals

Catalog number: A15547.09
10 g, Each
Thermo Scientific Chemicals

Chlorotriethylsilane, 98+%, Thermo Scientific Chemicals

Catalog number: A15547.09
10 g, Each
Quantity
Catalog number: A15547.09
also known as A15547-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
994-30-9
IUPAC Name
chlorotriethylsilane
Molecular Formula
C6H15ClSi
InChI Key
DCFKHNIGBAHNSS-UHFFFAOYSA-N
SMILES
CC[Si](Cl)(CC)CC
Assay (GC)
≥98.0% (non-U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
Clear colorless to pale yellow
Comment
Specification differs for U.S. and non-U.S. material where indicated
Form
Liquid

Description

Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

Solubility
Soluble in aprotic organic solvents.

Notes
Incompatible with bases, strong oxidizing agents, strong acids, alcohols, amines, ammonia, aldehydes and acetone.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text