Thermo Scientific Chemicals

5-Bromopentyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: A15549.22
100 g, Each
Thermo Scientific Chemicals

5-Bromopentyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: A15549.22
100 g, Each
Quantity
Catalog number: A15549.22
also known as A15549-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15848-22-3
IUPAC Name
5-bromopentyl acetate
Molecular Formula
C7H13BrO2
InChI Key
JIHJLQWUVZUKCH-UHFFFAOYSA-N
SMILES
CC(=O)OCCCCCBr
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4590-1.4630 @ 20°C

Description

5-?Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-​Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text