Thermo Scientific Chemicals

Lead(IV) acetate, 96% (dry wt.), stab. with 5-10% glacial acetic acid, Thermo Scientific Chemicals

Catalog number: A15551.22
100 g, Each
Thermo Scientific Chemicals

Lead(IV) acetate, 96% (dry wt.), stab. with 5-10% glacial acetic acid, Thermo Scientific Chemicals

Catalog number: A15551.22
100 g, Each
Quantity
Catalog number: A15551.22
also known as A15551-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
546-67-8
IUPAC Name
λ²-lead(2+) tetraacetate
Molecular Formula
C8H12O8Pb
InChI Key
ACKFDYCQCBEDNU-UHFFFAOYSA-J
SMILES
[Pb++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Comment
May contain fine dark particles from the manufacturing process
Form
Moist crystals or powder or crystalline powder
Appearance (Color)
White to pale cream or pale grey
Assay from Supplier's CofA
≥95.0% (ex Pb; dry wt. basis, Iodometry)

Description

Lead(IV) acetate is an important oxidizing agent and a source of acetyloxy group used in organic synthesis. For example, 1,4-dioxene is prepared from dioxane involving 2-acetoxy-1,4-dioxane as an intermediate. Similarly, it is used for the preparation of bis(trifluoromethyl)diazomethane from hexafluoroacetone hydrazone. It also reacts with alkenes, alcohols having a delta-proton and di-n-butyl d-tartrate to get gamma-lactones, cyclic ethers and n-butyl glyoxylate respectively. It induces the cleavage of 1,2-diols to the corresponding aldehydes or ketones. It is actively involved in the Kochi reaction for the decarboxylation of carboxylic acids to alkyl halides and used as an alternative reagent to bromine in the Hofmann rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(IV) acetate is an important oxidizing agent and a source of acetyloxy group used in organic synthesis. For example, 1,4-dioxene is prepared from dioxane involving 2-acetoxy-1,4-dioxane as an intermediate. Similarly, it is used for the preparation of bis(trifluoromethyl)diazomethane from hexafluoroacetone hydrazone. It also reacts with alkenes, alcohols having a delta-proton and di-n-butyl d-tartrate to get gamma-lactones, cyclic ethers and n-butyl glyoxylate respectively. It induces the cleavage of 1,2-diols to the corresponding aldehydes or ketones. It is actively involved in the Kochi reaction for the decarboxylation of carboxylic acids to alkyl halides and used as an alternative reagent to bromine in the Hofmann rearrangement.

Solubility
Soluble in water, ethanol, chloroform, benzene, nitrobenzene, tetrachloroethane, nitric acid, hot acetic acid and hydrochloric acid.

Notes
Air and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, strong acids and strong reducing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text