Thermo Scientific Chemicals

Thiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A15553.14
25 g, Each
Thermo Scientific Chemicals

Thiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A15553.14
25 g, Each
Quantity
Catalog number: A15553.14
also known as A15553-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16629-19-9
IUPAC Name
thiophene-2-sulfonyl chloride
Molecular Formula
C4H3ClO2S2
InChI Key
VNNLHYZDXIBHKZ-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=CS1
Form
Crystals or powder or crystalline powder or chunks or fused solid
Melting Point (clear melt)
26.0-35.0?C
Appearance (Color)
White to orange or blue to pale gray to gray to black
Assay (GC)
≥96.0%

Description

2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text