Thermo Scientific Chemicals

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Catalog number: A15554.0E
2500 g, Each
Thermo Scientific Chemicals

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Catalog number: A15554.0E
2500 g, Each
Quantity
Catalog number: A15554.0E
also known as A15554-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-39-5
IUPAC Name
ethyl 2-chloroacetate
Molecular Formula
C4H7ClO2
InChI Key
VEUUMBGHMNQHGO-UHFFFAOYSA-N
SMILES
CCOC(=O)CCl
Assay (GC)
≥98.5%
Refractive Index
1.4190-1.4230 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow

Description

Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

Solubility
Soluble in water 20 g/L (20°C).

Notes
Incompatible with acids, bases, oxidizing agents, reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text