Assay from Supplier's CofA≥96.0% (GC, US Sourced Material)
Melting Point (clear melt)136.0-145.0?C (UK Sourced Material)
FormPowder
CommentMaterial Sourced in UK and US
Appearance (Color)Orange
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.