Thermo Scientific Chemicals

4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A15557.14
25 g, Each
Thermo Scientific Chemicals

4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A15557.14
25 g, Each
Quantity
Catalog number: A15557.14
also known as A15557-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-51-2
IUPAC Name
4-methoxybenzene-1,2-diamine
Molecular Formula
C7H10N2O
InChI Key
AGAHETWGCFCMDK-UHFFFAOYSA-N
SMILES
COC1=CC=C(N)C(N)=C1
Appearance (Color)
Grey or brown or purple
Assay (GC)
>97.5%
Form
crystalline or fused solid

Description

4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as a derivatizing reagent by High-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as a derivatizing reagent by High-performance liquid chromatography (HPLC).

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Air sensitive and Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text