Thermo Scientific Chemicals

4-Methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A15559.0E
2500 g, Each
Thermo Scientific Chemicals

4-Methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A15559.0E
2500 g, Each
Quantity
Catalog number: A15559.0E
also known as A15559-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-13-5
IUPAC Name
(4-methoxyphenyl)methanol
Molecular Formula
C8H10O2
InChI Key
MSHFRERJPWKJFX-UHFFFAOYSA-N
SMILES
COC1=CC=C(CO)C=C1
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.5430-1.5470 @ 20?C
Appearance (Color)
Clear colorless to yellow

Description

4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

Solubility
Soluble in water, chloroform, ethyl acetate.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text