Thermo Scientific Chemicals

Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals

Catalog number: A15568.22
100 g, Each
Thermo Scientific Chemicals

Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals

Catalog number: A15568.22
100 g, Each
Quantity
Catalog number: A15568.22
also known as A15568-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
582-52-5
IUPAC Name
5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular Formula
C12H20O6
InChI Key
KEJGAYKWRDILTF-UHFFFAOYNA-N
SMILES
CC1(C)OCC(O1)C1OC2OC(C)(C)OC2C1O
Form
Crystalline powder
Optical Rotation
-11? ± 1? (c=5 in Ethanol)
Assay (GC)
>98.0%
Solution Test
Clear (5% in boiling Water)
Appearance (Color)
White to pale brown

Description

It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

Solubility
Solubility in hot water gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text