Thermo Scientific Chemicals

Iron(II) sulfide, tech., Thermo Scientific Chemicals

Catalog number: A15569.0B
1000 g, Each
Thermo Scientific Chemicals

Iron(II) sulfide, tech., Thermo Scientific Chemicals

Catalog number: A15569.0B
1000 g, Each
Quantity
Catalog number: A15569.0B
also known as A15569-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-37-9
IUPAC Name
λ²-iron(2+) sulfanediide
Molecular Formula
FeS
InChI Key
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES
[S--].[Fe++]
Form
Powder and/or Chunks
Comment
Volatile matter: <0.07% (@ 105?C)
Appearance (Color)
Grey to black
Comment
Sulfide test = Postive
Particle Size
Sieve residue <13.5% +325 mesh (44 micron)

Description

Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

Solubility
Soluble in water(0.0062g/L ), in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.

Notes
Moisture Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agent, acids, air, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text