Thermo Scientific Chemicals

Pentafluorophenol, 99%, Thermo Scientific Chemicals

Catalog number: A15574.22
100 g, Each
Thermo Scientific Chemicals

Pentafluorophenol, 99%, Thermo Scientific Chemicals

Catalog number: A15574.22
100 g, Each
Quantity
Catalog number: A15574.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
771-61-9
Appearance (Color)
White to pale cream
Form
Crystalline or fused solid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

Pentafluorophenol is used in the preparation of pentafluorophenyl esters for peptide synthesis and vulcanized polymers. It is also used for peptide coupling and heterocyclic acid derivatives. It is involved in the preparation of pentafluorophenyl formate which acts as a formulating agent for amines and amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenol is used in the preparation of pentafluorophenyl esters for peptide synthesis and vulcanized polymers. It is also used for peptide coupling and heterocyclic acid derivatives. It is involved in the preparation of pentafluorophenyl formate which acts as a formulating agent for amines and amino acids.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, bases, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text