Thermo Scientific Chemicals

2-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15576.22
100 g, Each
Thermo Scientific Chemicals

2-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15576.22
100 g, Each
Quantity
Catalog number: A15576.22
also known as A15576-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70-11-1
IUPAC Name
2-bromo-1-phenylethan-1-one
Molecular Formula
C8H7BrO
InChI Key
LIGACIXOYTUXAW-UHFFFAOYSA-N
SMILES
BrCC(=O)C1=CC=CC=C1
Appearance (Color)
White or pale yellow to pale green or pale brown or green-brown
Form
Crystalline powder or crystals
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
46-52°C

Description

It is used in preparation of crystalline esters from acids. 2-Bromoacetophenone is a complete and irreversible inactivator of human liver aldehyde dehydrogenase isoenzymes E1 and E2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of crystalline esters from acids. 2-Bromoacetophenone is a complete and irreversible inactivator of human liver aldehyde dehydrogenase isoenzymes E1 and E2.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids & bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text